Value Plastics® Adapter Fittings, Luer to Luer, Polycarbonate
Supplier: VALUE PLASTICS TE
FTLLC-91000 FTLLP-91000 FTLT-91000 LC78-91000 LE87-91000 MTLLP-91000
MFLX45501-22BLKEA 571.96 CAD
MFLX45501-22BLK MFLX45501-28BLK MFLX45501-56BLK MFLX45501-82BLK MFLX45501-84BLK MFLX45504-56BLK
Value Plastics® Adapter Fittings, Luer to Luer, Polycarbonate
Fittings and Connectors
Easily transition to a luer fitting.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Easily complete your luer fittings systems with these premium connectors. Included are some special configurations available only in polycarbonate. All female ports may be used with either slip or luer lock connections. Plugs and caps are also available. Polycarbonate material has good chemical compatibility and can be sterilized via ethylene oxide or gamma irradiation.