Value Plastics® Adapter Fittings, Luer to Luer, Nylon
Supplier: VALUE PLASTICS TE
FTLLP-11000 FTLLC-11000 FTLT-11000 LC34-11000 LE87-11000 LT878-11000 MTLLP-11000 MTLCS-11000
MFLX45502-28BLKEA 406.06 CAD
MFLX45502-28BLK MFLX45502-22BLK MFLX45502-56BLK MFLX45502-80BLK MFLX45502-84BLK MFLX45502-85BLK MFLX45505-56BLK MFLX45505-72BLK
Value Plastics® Adapter Fittings, Luer to Luer, Nylon
Fittings and Connectors
Easily transition to a luer fitting.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery