Value Plastics® Adapter Fittings, Female Luer to Threaded, Straight
Supplier: VWR International
MFLX30800-00BLK MFLX45508-76BLK KFTLL-91000 SFTLL-91000 KFTLL-6005 SFTLL-60051000
MFLX30800-00BLKEA 1510.48 CAD
MFLX30800-00BLK MFLX45508-76BLK MFLX45501-60BLK MFLX45501-66BLK MFLX45508-60BLK MFLX45508-66BLK
Value Plastics® Adapter Fittings, Female Luer to Threaded, Straight
Fittings and Connectors
Easily transition between luer fittings and UNF or NPT threads.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
These adapters transition between luer fittings and UNF or NPT threads. Both male and female luer connections are available. All threaded connections are female thread.