Smiths Medical® Luer Fittings, High-Pressure Stopcock
Supplier: Smiths Medical
B11431LM (200PK)
MFLX30526-34BLKEA 850.04 CAD
MFLX30526-34BLK
Smiths Medical® Luer Fittings, High-Pressure Stopcock
Valves Ball Valves
Pressure rating up to 1050 psi.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Stopcock fittings feature luer connections.