Smiths Medical® Luer Fittings on PVC Flexible Tubing, Straight
Supplier: ICU MEDICAL, INC.
B1691 (100PK) B1692 (100PK) B1694 (100PK) B1562 (100PK) B1572 (100PK)
MFLX30526-14BLKEA 292.3 CAD
MFLX30526-14BLK MFLX30526-16BLK MFLX30526-18BLK MFLX30600-60BLK MFLX30600-62BLK
Smiths Medical® Luer Fittings on PVC Flexible Tubing, Straight
Fittings and Connectors
Get a seamless transition from source to system.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
This high pressure flexible PVC tubing has luer fittings at each end and comes in various lengths. Can be sterilized by ETO.