Select Acid Rinse, StatLab
Supplier: StatLab
SL404
76548-236EA 90.8 CAD
76548-236
Select Acid Rinse, StatLab
Cleaning Agents Detergents and Surface Cleansers
Produces unparalleled crisp nuclear chromatin detail and eliminates undesirable background staining. Utilizes a buffer system to prevent pH fluctuation.
  • Premixed and ready-to-use
  • Non-hazardous
Packaging: Manufactured in the USA