Diethyl ether, EMPROVE® ESSENTIAL Ph. Eur., BP, SAFC®
Supplier: MilliporeSigma
|
Danger
|
Stabilization:
Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 5 - 10 ppm
M-Clarity™ quality level = MQ500 (more info.)
1.00926.1000
1.00926.6025
CA1.00926.1000CS
1105.9
CAD
CA1.00926.1000
CA1.00926.6025
Diethyl ether, EMPROVE® ESSENTIAL Ph. Eur., BP, SAFC®
Diethyl ether
Formula:
(CH₃CH₂)₂O Boiling Pt: 35 °C (1013 hPa) Melting Pt: –123 °C Density: 0.7135 g/cm³ (20 °C) Flash Pt: –40 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011646 CAS Number: 60-29-7 EINECS: 200-467-2 UN: 1155 ADR: 3,I REACH: 01-2119535785-29 Merck Index: 12,03852 |
|
Specification Test Results
Purity (GC) | ≥ 99.5 % |
Identity (IR) | conforms |
Acidic substances | conforms |
Density (d 20 °C/20 °C) | 0.714 - 0.716 |
Boiling point (lower value) | ≥ 34.0 °C |
Boiling point (upper value) | ≤ 35.0 °C |
Foreign odor | conforms |
Aldehydes | conforms |
Peroxides | conforms |
Residual solvents (ICH Q3C) | excluded by production process |
Evaporation residue | ≤ 20 mg/l |
Water | ≤ 0.20 % |
Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*). |
|
Stabilized with 5 - 10 ppm 2,6-Di-tert-butyl-4-methylphenol (BHT). | |
Conforms to Ph Eur, BP |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...