Essentials Powder Funnels, Antylia Scientific
Supplier: Antyila Scientific
06286-53 06286-54 06286-55 06286-56
76533-752PK 185.99 CAD
76533-752 76533-754 76533-756 76533-758
Essentials Powder Funnels, Antylia Scientific
Funnels Powder Funnels
Prevent blockages with wide base opening.
  • Constructed from durable food grade plastic
  • Cleanroom class 100,000 packed and individually bagged
  • Disposable funnels are ideal for food and pharmaceutical applications
These essentials powder funnels are ideal for transferring powders and granules. Disposable funnels are packaged in class 100,000 cleanroom and individually bagged to avoid cross contamination. The wide opening at the base of the funnel will help prevent powder blockages.