Essentials LiquiThief Liquid Sampler, HDPE, Antylia Scientific
Supplier: Antyila Scientific
06286-57 06286-58 06286-59 06286-60
76533-760PK 406.81 CAD
76533-760 76533-762 76533-764 76533-766
Essentials LiquiThief Liquid Sampler, HDPE, Antylia Scientific
Samplers Fluid Samplers
Reduce cross contamination while sampling low viscosity liquids and oils.
  • Manufactured in Class 100,000 cleanroom and individually packaged
  • Constructed of chemically inert high-density polyethylene (HDPE)
The LiquiThief liquid samplers are designed to easily take liquid samples with the added convenience of not having to clean the sampler prior to use. The samplers are manufactured, assembled, and packed in a Class 100,000 cleanroom to ensure cleanliness and they are individually bagged. Choose from sterile or non sterile samplers. Sterile samplersMore Product Information