Essentials PowderThief, Sterile HDPE, Antylia Scientific
Supplier: Antyila Scientific
06286-30 06286-31 06286-32 06286-33
76533-714PK 509.68 CAD
76533-714 76533-716 76533-718 76533-720
Essentials PowderThief, Sterile HDPE, Antylia Scientific
Samplers Solid Samplers
Quickly and easily take a sample at a specified point within the vessel.
  • Manufactured in Class 100,000 cleanroom and individually packaged
  • Constructed of chemically inert high-density polyethylene (HDPE)
  • Sterile
These PowderThief powder samplers are designed to take representative samples from known locations within the bulk powder. The samplers are manufactured, assembled, and packed in a Class 100,000 cleanroom to ensure cleanliness and they are individually bagged. Samplers are also sterilized via gamma irradiation (minimum dose 25 kGy).