Essentials Sleeve Sample Chamber and Handle, Antylia Scientific
Supplier: Antyila Scientific
06286-15 06286-16
76533-698EA 595.05 CAD
76533-698 76533-700
Essentials Sleeve Sample Chamber and Handle, Antylia Scientific
Samplers Fluid Samplers
Reliably sample large volumes at great depths.
  • Designed to be extremely strong and robust to allow samples to be taken from depths up to 6 meters
  • Constructed of durable 316L stainless steel
  • Non sterile
The sleeve sampler is ideal for sampling large volumes of powder or granules at a depth of up to 6 meters. The heavy duty 316L stainless steel handle provides enhanced strength and durability. Simply push the sleeve sampler into the powder or granules to the required depth and pull up on the handle to open the sleeve and take a sample. Sleeve samplerMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable