Essentials Grain Spears, Antylia Scientific
Supplier: Antyila Scientific
06286-13 06286-14
76533-694EA 751.86 CAD
76533-694 76533-696
Essentials Grain Spears, Antylia Scientific
Samplers Fluid Samplers
Reliably sample various powders, granules, and grains.
  • Suitable for sampling from all layers of the container, providing an accurate representation of contents
  • Constructed of lightweight aluminum for ease of use and durability
These grain samplers provide highly representative sampling of powders, granules, and grains. The spiral position of the pockets means the bottom pocket opens and fills first, giving a more representative sample. The lightweight aluminum construction means even the long samplers are easy to operate and manage. The tip is fixed permanently so it cannot be lost. The sampler is manufactured so that it can be stripped and cleaned within seconds. Sampler handle sold separately.