Essentials Powered Ice Samplers, 316L Stainless Steel, Antylia Scientific
Supplier: Antyila Scientific
06286-01 06286-02
76533-670EA 526.08 CAD
76533-670 76533-672
Essentials Powered Ice Samplers, 316L Stainless Steel, Antylia Scientific
Samplers Solid Samplers
Ideal sampling tool for wax and frozen products.
  • Designed to fit most standard electric drills
  • High-quality 316L stainless steel construction
Quickly cut through hard, frozen samples by attaching the ice sampler to an electric hand drill. Once powered on, the sharp-edged sampler rotates and cuts the material into the sampler. Crevice-free, highly polished surface sampler is easy to clean.
Certifications: Supplier certificate of conformity is included with product shipment.