Essentials Powder Lances, HDPE, Antylia Scientific
Supplier: Antyila Scientific
06286-41 06286-42 06286-43 06286-44 06286-45 06286-46
76533-728PK 477.87 CAD
76533-728 76533-730 76533-732 76533-734 76533-736 76533-738
Essentials Powder Lances, HDPE, Antylia Scientific
Samplers Solid Samplers
Quickly and easily take a cross sectional sample within the vessel.
  • Manufactured in Class 100,000 cleanroom and individually packaged
  • Ergonomically designed handle makes it easy to use, even when doing a large amount of sampling
  • Constructed of chemically inert high-density polyethylene (HDPE)
These powder lance samplers are designed to take a cross sectional sample at an exact position within the vessel. The samplers are manufactured, assembled, and packed in a Class 100,000 Cleanroom to ensure cleanliness and individually bagged. Choose from sterileMore Product Information