Essentials Industrial Scoops, Stainless Steel, Antylia Scientific
Supplier: Antyila Scientific
06700-63 06700-64 06700-65 06700-66 06700-67
76533-768EA 42.24 CAD
76533-768 76533-770 76533-772 76533-774 76533-776
Essentials Industrial Scoops, Stainless Steel, Antylia Scientific
Scoops
Fully welded handle design provides enhanced durability.
  • Constructed of durable stainless steel
  • Smooth surface ideal for food sampling
  • High quality mirror polish allows for easy cleaning
These stainless steel industrial scoops feature a fully welded handle for optimal durability in rugged applications. Each scoop is constructed of stainless steel with a surface finish better than 0.5 µm Ra, allowing for quick cleaning. These scoops come with an angled handle for easy scooping.