Cleanroom Papers, Columbia Cleanroom
Supplier: Columbia Printing & Graphics
PA22LF-SAL-BAR11 PA22LF-SAL-BOR11 PA28LF-BLU-BAR11 PA28LF-BLU-BOR11 PA28LF-CAN-BAR11 PA28LF-CAN-BOR11 PA28LF-GRE-BAR11 PA28LF-GRE-BOR11 PA28LF-ORCH-BAR11 PA22LF-BLU-BAR11 PA22LF-BLU-BOR11 PA22LF-CAN-BAR11 PA22LF-CAN-BOR11 PA22LF-GRE-BAR11 PA22LF-GRE-BOR11 PA22LF-ORCH-BAR11 PA22LF-ORCH-BOR11 PA28LF-ORCH-BOR11 PA28LF-SAL-BAR11 PA28LF-SAL-BOR11
76635-230CS 323.56 CAD
76635-230 76635-250 76635-232 76635-252 76635-234 76635-254 76635-236 76635-256 76635-238 76635-222 76635-242 76635-224 76635-244 76635-226 76635-246 76635-228 76635-248 76635-258 76635-240 76635-260
Cleanroom Papers, Columbia Cleanroom
Paper
White, 8.5×11" cleanroom paper with colored bar or edge to distinguish between departments, operations or projects.
  • Class 10
  • No chance for ink bleeds
  • Latex-free
  • Cellulose and low-density polyethylene co-polomers
Packaging: Double-bagged in packs of 250 sheets. 10 packs of 250 sheets/case.