Coriolis® Air Sampler Accessories, Bertin Instruments
Supplier: BERTIN CORP
S001109-CORM0-A.0 S001111-CORM0-A.0 S001112-CORM0-A.0 S001113-CORM0-A.0
76636-430PK 503.42 CAD
76636-430 76636-432 76636-434 76636-436 76636-438 76636-440 76636-450
Coriolis® Air Sampler Accessories, Bertin Instruments
Samplers Air Samplers
Coriolis® air sampler consumables are part of the innovative dry cyclonic technology, which aspirates airborne particles and centrifuges them in a cone.
  • Single use consumables
  • Ensure optimal collection efficiency for Coriolis® air samplers
The separation of the airborne particles from the air flow is due to the air flow rate, the air intake geometry, the design of the cones, and the collection liquid (surfactant in low concentration). The collected particles or microorganisms can then be recovered by rinsing the consumables with an appropriate bufferMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable