Polycarbin® Racked LTS® Compatible Pipette Tips
Supplier: POLYCARBIN, INC.
PC02-04-05 PC01-04-06 PC02-05-05 PC01-05-06 PC02-06-05 PC01-06-06
76635-492CS 921.71 CAD
76635-492 76635-490 76635-488 76635-498 76635-484 76635-508 76635-482
Polycarbin® Racked LTS® Compatible Pipette Tips
Pipette Tips
Polycarbin’s pipette tip rack is made of 100% circular economy, post-consumer recycled content, combining high performance with a significantly reduced carbon footprint (tips do not contain recycled content).
  • Rainin LTS® compatible
  • Carries a smaller carbon footprint
  • DNase, RNase and pyrogen free
  • Sterile
Polycarbin's line of low-carbon lab products are designed for the sustainability-minded scientist. When designing our line of pipette tips, we focused on the the tip box, or 'rack,' which is the most dense, carbon-intensive piece of plastic. All tipMore Product Information