Evacuable KBr Pellet Dies, Firebird Optics
Supplier: FIREBIRD OPTICS
KB-10 KB-13 KB-16 KBX-20 KBX-25 KBX-32 KBX-35 KBX-40 KBX-45 KB-3 KB-5 KB-6 KB-7 KB-8
76636-338EA 1859.25 CAD
76636-338 76636-340 76636-344 76636-382 76636-384 76636-386 76636-388 76636-390 76636-392 76636-348 76636-350 76636-352 76636-354 76636-356
Evacuable KBr Pellet Dies, Firebird Optics
Presses
Dies suitable for producing discs of 3, 5, 6, 7, 8, 10, 13, 16, 20, 25, 32, 35 and 40 mm diameter as available as standard within the size range for the KB series. Non-standard sizes are available on request.
  • All dies in the KB series are constructed entirely of stainless tool steel, and parts are precision machined
  • Pellets are optically flat and mirror polished
  • Tungsten carbide pellets and component parts can be supplied to special order
  • Dies are supplied with black anodised sleeves
KBr Disc Technique: This widely practiced method consists ofMore Product Information