Bioquell BQ-50 Hydrogen Peroxide Vapor Generator
Supplier: Bioquell
VWRBQ50
76635-458EA 171635.38 CAD
76635-458
Bioquell BQ-50 Hydrogen Peroxide Vapor Generator
Disinfection Systems
A mobile and robust hydrogen peroxide vapor generator, the Bioquell BQ-50, when utilized with Bioquell Hydrogen Peroxide Sterilant (EPA Reg. No. 72372-1-86703) can be used hospital-wide to disinfect exposed surfaces. Get automated and proven results that can support in your infection control plans with Bioquell.
  • Mobile bio-decontamination system ideal for critical care areas, operating rooms, patient rooms, emergency response preparedness and more
  • 6-log sporicidal kill using Bioquell Hydrogen Peroxide Sterilant (EPA Registration Number: 72372-1-86703)
  • Automated cycles after entering in room dimensions (up to 200m3 / 7063 ft3)
  • RFID bottle reading for safety and quality control
  • No wipe downs or additional steps required after cycle completion
  • Requires only a single cycle
  • Can bio-decontaminate connected adjacent bathroom
  • Options for using multiple systems in high volume areas
  • Comprehensive safety and user training
A mobile and robust hydrogen peroxide vapor generator, the Bioquell BQ-50 can be used hospital-wide. The technology, when utilized with Bioquell Hydrogen Peroxide Sterilant, kills microorganisms from exposed surfaces. Get automated and proven results that can provide assistance in your infection control plans.
The Bioquell BQ-50 helps eliminate microorganisms in nearly every location within a facility. Facilities can work to reduce the environment's impact as a source of infection by bio-decontaminating areas where patients were recently treated or will be receiving treatment, such as prior to admitting immuno-compromised patients or in rooms previously occupied by a person on contact precautions. From Neonatal and Oncology to the Operating Room, using Bioquell in the right rooms at the right time kills microorganisms that otherwise patients would have potentially been exposed to. Note: This product is not for use as a sterilant on surgical instruments.