RayDyLyo® Sterile Snap Caps, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
255855003 255845001 255845002 255845003 255853000 255853001 255853002 255853003 255854001 255854004 255855001 255843000 255843001 255843002 255843003 255845000
76632-802CS 18615.62 CAD
76632-802 76632-786 76632-780 76632-788 76632-820 76632-810 76632-830 76632-782 76632-794 76632-818 76632-784 76632-826 76632-790 76632-828 76632-808 76632-824
RayDyLyo® Sterile Snap Caps, DWK Life Sciences
Closures Chromatography Vial Closures
RayDyLyo® plastic push-fit caps.
  • Flip off snap caps are pre-assembled with an ISO 8362-2 compliant elastomer stopper
  • Container closure integrity (CCI) validated for use with ISO 8362-1 glass and COP vials
  • Delivered sterile (RTU) with a 2-year shelf life from time of manufacturer
  • Clean room manufactured with 100% in-process quality controls
  • Suitable for aseptic small batch, clinical trial, and robotic fill and finish operations
  • Snap caps do not require special crimping tools and affix with single vertical pressure
  • Provides tamper-evident closing and eliminates crimping defects
  • Compatible with gamma and steam terminal sterilization
  • Available in nested, standard bag, and RTP-type bag configurations
RayDyLyo® plastic push-fit caps from DWK Life Sciences, designed for closing vials of injectable drugs. These flip-off snap caps are pre-assembled with an ISO 8362-2 compliant elastomer stopper to simplify the production process.