EZ-fill® Vials, Stevanato Group
Supplier: DWK Life Sciences (KIMBLE)
P05B2003.6434 P05B2526.6434 P05B2526.6492 P06B2001.6461 P06B2527.6461 05B2003.6492 05B2060.6924 05B2536.6924 06B2526.7082 06B2543.7082 06B2020.6461
76632-792CS 6208.81 CAD
76632-792 76632-796 76632-806 76632-816 76632-798 76632-814 76632-822 76632-812 76632-804 76632-800 77020-362
EZ-fill® Vials, Stevanato Group
Vials
EZ-fill® Vials, offered in nest and tub and trays.
  • Clean, depyrogenated, and sterilized ready-to-use (RTU) vials designed for pharmaceutical and contract manufacturing entities performing small to large batch aseptic parenteral packaging
  • ISO 8362-1 2R, 4R, 6R, 10R, and 20R vials, manufactured from borosilicate type 1, 51 expansion glass, with European Blowback
  • Nested configurations are designed for direct entry into flex fill, robotic, and combi filling operations
  • Tray configurations are suitable for small batch and in-line filling operations
  • Reduces total cost of ownership and time to market
EZ-fill® vials are offered in RTU nest and tub and tray configurations eliminating glass-to-glass contact to assure pristine cosmetic quality. The pre-sterilized formats are designed for direct entry into your aseptic fill operation allowing you to maximize efficiencies and to minimize total cost.