Pro-Wipe® SuperSorb Wipes, Berkshire
Supplier: Berkshire
PWSS0914D16 PWSS1114QF18
76595-250CS 440.77 CAD
76595-250 76595-254
Pro-Wipe® SuperSorb Wipes, Berkshire
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Durable and low-linting Pro-Wipe® SuperSorb wipes are manufactured from 80% cellulose/10% lyocell/10% bicomponent.
  • No binders or other chemical additives
  • Smooth and soft with high strength and durability
  • Knife cut edges
  • Highly absorbent
  • Good wet strength
  • Chemically compatible with common cleaning and disinfecting solutions
  • Absorbs water and oil based solutions
Pro-Wipe® SuperSorb wipes are medium-duty, all-purpose industrial nonwoven wipes comprised of cellulose/lyocell/bicomponent materials. They are durable and low-linting. Pro-Wipe® Supersorb wipes are tough enough to be a rag replacement and clean enough to be used in a controlled manufacturing environment.
Packaging: 11×14" wipes are stacked ¹/₄ fold, 50 sheets/pack, 18 packs/case. 9×14" wipes are packed C-fold in a dispenser box, 60 sheets/box, 10 boxes/case.