VWR® Wide Bore Pipette Tips
Supplier: VWR International
76635-670 76635-646 76635-650 76635-632 76635-638 76635-658 76635-652 76635-634
76635-670CS 256.12 CAD
76635-670 76635-646 76635-650 76635-632 76635-638 76635-658 76635-652 76635-634
VWR® Wide Bore Pipette Tips
Pipette Tips
Larger orifice prevents cell fragmentation due to shearing.
  • Ideal for NGS protocol
Ideal for pipetting of macromolecules and viscous samples.
Certifications: Certified free of detectable RNase, DNase, DNA, PCR inhibitors, and tested pyrogen-free. Sterile product is certified pyrogen-free.