VWR® Cleanroom Gloves Class 10 Nitrile Non-Tacky Gloves
Supplier: VWR International
76533-194 76533-196 76533-198 76533-200 76533-202 76533-204
76533-194CS 396.26 CAD
76533-194 76533-196 76533-198 76533-200 76533-202 76533-204
VWR® Cleanroom Gloves Class 10 Nitrile Non-Tacky Gloves
Gloves
VWR® Cleanroom nitrile gloves are engineered for superior protection and comfort in controlled environments. These non-tacky gloves meet stringent Class 10 (ISO 4) standards, ensuring minimal particulate contamination. Ideal for pharmaceutical, biotech, and electronics industries, they offer excellent chemical resistance and tactile sensitivity for critical cleanroom tasks.
  • Class 10 / ISO 4
  • Nitrile
  • Silicone-free
  • Beaded cuff
  • Low particle count
  • ESD performance: 10⁹ ohms (CAFÉ), 10¹¹ ohms (Surface), 10⁹ ohms (Volume)
VWR® Cleanroom non-tacky nitrile gloves are designed for critical environments, including pharmaceutical, biotech, and electronics industries. Compliant with ISO 4 (Class 10) standards, they minimize contamination risks. These non-tacky gloves ensure superior particulate control and tactile sensitivity, making them ideal for precision applications requiring clean, controlled handling.
Packaging:
Each bag contains 100 cleanroom gloves, ensuring optimal hygiene and contamination control. The gloves are packaged in a cleanroom environment, maintaining  integrity. Each case includes 10 bags, totaling 1000 gloves per case. This packaging design allows for easy access and storage while ensuring that all gloves remain clean and uncontaminated until ready for use.