VWR® Macro Pipette Tips
Supplier: VWR International
76635-674 76635-672 76635-648 76635-642 76635-664 76635-644 76635-660 76635-640 76635-666 76635-662 76635-656 76635-630 76635-628 76635-668
76635-674CS 278.38 CAD
76635-674 76635-672 76635-648 76635-642 76635-664 76635-644 76635-660 76635-640 76635-666 76635-662 76635-656 76635-630 76635-628 76635-668
VWR® Macro Pipette Tips
Pipette Tips
Improved accuracy over serological pipettes.
  • Tip length facilitates access into bottles, cell culture flasks, and tubes
Universal fit for compatibility with most major pipettor brands.
Certifications: Certified free of detectable RNase, DNase, DNA, PCR inhibitors, and tested pyrogen-free. Sterile product is certified pyrogen-free.