cis-​Ethyl 4-​aminocyclohexanecarb​oxylate HCl salt 95%

Supplier: AFG Bioscience

C78593-1G C78593-5G
76620-408EA 395.6 CAD
76620-408 76620-410
cis-​Ethyl 4-​aminocyclohexanecarb​oxylate HCl salt 95%
cis-​Ethyl 4-​aminocyclohexanecarb​oxylate HCl salt
Formula: C₉ClH₁₈NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD01333987
CAS Number: 3685-28-7

Order Now

Specification Test Results

Purity Limit 95%
Appearance White Solid

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR