(2S)-2-Decanamido-3-methylbutanoic acid 95%
Supplier: AFG BIOSCIENCE LLC
C73078-100MG
C73078-200MG
76620-404EA
712.09
CAD
76620-404
76620-406
(2S)-2-Decanamido-3-methylbutanoic acid 95%
(2S)-2-Decanamido-3-methylbutanoic acid
Formula:
C₁₅H₂₉NO₃ MW: 271.4 g/mol Storage Temperature: Refrigerator |
CAS Number:
22220-07-1 |
Specification Test Results
Purity Limit | 95% |
Appearance | white powder |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...