3-Hydrazino-benzoic acid methyl ester HCl salt 95%

Supplier: AFG Bioscience

C72375-1G C72375-2G
76620-400EA 545.18 CAD
76620-400 76620-402
3-Hydrazino-benzoic acid methyl ester HCl salt 95%
3-Hydrazino-benzoic acid methyl ester HCl salt
Formula: C₈ClH₁₁N₂O₂
Storage Temperature: Ambient
CAS Number: 473575-39-2

Order Now

Specification Test Results

Purity Limit 95%
Appearance Yellow Solid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR