(R)-O-ISOPROPYLIDENE GLYCEROL mesylate 95%
Supplier: AFG Bioscience
C71340-1G
C71340-5G
76620-396EA
389.41
CAD
76620-396
76620-398
(R)-O-ISOPROPYLIDENE GLYCEROL mesylate 95%
(R)-O-ISOPROPYLIDENE GLYCEROL mesylate
Formula:
C₇H₁₄O₅S Storage Temperature: Refrigerator |
CAS Number:
83461-40-9 |
Specification Test Results
Purity Limit | 95% |
Appearance | Yellow Oil |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...