methyl 2-(2-chlorophenoxy)propanoate 95%

Supplier: AFG Bioscience

C71038-5G C71038-1G
76620-394EA 1186.8 CAD
76620-394 76620-392
methyl 2-(2-chlorophenoxy)propanoate 95%
methyl 2-(2-chlorophenoxy)propanoate
Formula: C₁₀H₁₁ClO₃
Storage Temperature: Ambient
CAS Number: 122623-80-7

Order Now

Specification Test Results

Purity Limit 95%
Appearance Light Yellow Oil


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR