Tos-PEG7-t-butyl ester 95%

Supplier: AFG Bioscience

Synonyms: Tos-peg7-t-butyl ester, Tos-PEG6-C2-Boc

C59970-1G C59970-500MG C59970-5G
76620-388EA 545.18 CAD
76620-388 76620-386 76620-390
Tos-PEG7-t-butyl ester 95%
Tos-PEG7-t-butyl ester
Formula: C₂₆H₄₄O₁₁S
MW: 564.686 g/mol
Storage Temperature: Freezer
MDL Number: MFCD20926384
CAS Number: 406213-75-0

Order Now

Specification Test Results

Purity Limit 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR