DNP-​PEG3-​amine ≥95%

Supplier: AFG Bioscience

C59832-1G C59832-2G
76620-382EA 870.33 CAD
76620-382 76620-384
DNP-​PEG3-​amine ≥95%
DNP-​PEG3-​amine
Formula: C₁₂H₁₈N₄O₆
MW: 314.29 g/mol
Storage Temperature: Refrigerator
CAS Number: 929521-54-0

Order Now

Specification Test Results

Purity Limit > 95%
Appearance sticky oil


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR