DNP-​PEG4-​amine ≥95%

Supplier: AFG Bioscience

C59795-1G C59795-2G
76620-378EA 870.33 CAD
76620-378 76620-380
DNP-​PEG4-​amine ≥95%
DNP-​PEG4-​amine
Formula: C₁₄H₂₂N₄O₇
Storage Temperature: Refrigerator
CAS Number: 132316-46-2

Order Now

Specification Test Results

Purity Limit > 95%
Appearance sticky oil


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR