Acetamide, N-​[2-​[2-​(2-​aminoethoxy)​ethoxy]​ethyl]​-​2-​bromo-TFA salt 95%

Supplier: AFG Bioscience

C59525-1G C59525-250MG C59525-500MG
76620-366EA 623.06 CAD
76620-366 76620-362 76620-364
Acetamide, N-​[2-​[2-​(2-​aminoethoxy)​ethoxy]​ethyl]​-​2-​bromo-TFA salt 95%
Acetamide, N-​[2-​[2-​(2-​aminoethoxy)​ethoxy]​ethyl]​-​2-​bromo-TFA salt
Formula: C₈H₁₇BrN₂O₃·CF₃CO₂H
Storage Temperature: Refrigerator
CAS Number: 712279-20-4

Order Now

Specification Test Results

Purity Limit 95%
Appearance Yellow oil


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR