Thiol-PEG6-t-butyl ester
Supplier: AFG Bioscience
Synonyms:
HS-PEG6-CH2CH2COOtBu, HS-PEG6-CH2CH2-Boc
C59460-100MG
C59460-250MG
C59460-500MG
76620-356EA
389.41
CAD
76620-356
76620-358
76620-360
Thiol-PEG6-t-butyl ester
Thiol-PEG6-t-butyl ester
Formula:
C₁₉H₃₈O₈S MW: 426.565 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD26127815 CAS Number: 1818294-40-4 |
Specification Test Results
Appearance | Colorless Oil |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...