2-Fluoro-5-maleimidobenzoic acid ≥95%

Supplier: AFG Bioscience

C59219-1G C59219-2G
76620-346EA 791.2 CAD
76620-346 76620-348
2-Fluoro-5-maleimidobenzoic acid ≥95%
2-Fluoro-5-maleimidobenzoic acid
Formula: C₁₁H₆FNO₄
Storage Temperature: Refrigerator
CAS Number: 777920-02-2

Order Now

Specification Test Results

Purity Limit > 95%
Appearance Light yellow solid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR