1-iodo-​2-​{2-​[2-​(2-​iodoethoxy)​ethoxy]​ethoxy}​ethane 95%

Supplier: AFG Bioscience

C58967-10G C58967-1G C58967-5G
76620-344EA 1503.29 CAD
76620-344 76620-340 76620-342
1-iodo-​2-​{2-​[2-​(2-​iodoethoxy)​ethoxy]​ethoxy}​ethane 95%
1-iodo-​2-​{2-​[2-​(2-​iodoethoxy)​ethoxy]​ethoxy}​ethane
Formula: C₈H₁₆I₂O₃
Storage Temperature: Refrigerator
CAS Number: 36839-56-2

Order Now

Specification Test Results

Purity Limit 95%
Appearance Orange Oil


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR