1-[6-(aminooxy)hexyl]-1H-Pyrrole-2,5-dione hydrochloride ≥95%

Supplier: AFG Bioscience

C58765-100MG C58765-250MG C58765-500MG
76620-334EA 389.41 CAD
76620-334 76620-336 76620-338
1-[6-(aminooxy)hexyl]-1H-Pyrrole-2,5-dione hydrochloride ≥95%
1-[6-(aminooxy)hexyl]-1H-Pyrrole-2,5-dione hydrochloride
Formula: C₁₀H₁₆N₂O₃·xHCl
Storage Temperature: Refrigerator
CAS Number: 937025-28-0

Order Now

Specification Test Results

Purity Limit > 95%
Appearance white solid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR