N-(trifluoroacetyl)glycylGlycine NHS ester ≥95%
Supplier: AFG Bioscience
C57938-1G
C57938-2G
76620-330EA
778.81
CAD
76620-330
76620-332
N-(trifluoroacetyl)glycylGlycine NHS ester ≥95%
N-(trifluoroacetyl)glycylGlycine NHS ester
Formula:
C₁₀H₁₀F₃N₃O₆ MW: 325.2 g/mol Storage Temperature: Refrigerator |
CAS Number:
1401353-42-1 |
Specification Test Results
Purity Limit | > 95% |
Appearance | white solid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...