tert-​butyl 5-​hydroxypentanoate 95%

Supplier: AFG Bioscience

C57267-250MG C57267-500MG
76620-326EA 233.65 CAD
76620-326 76620-328
tert-​butyl 5-​hydroxypentanoate 95%
tert-​butyl 5-​hydroxypentanoate
Formula: C₉H₁₈O₃
Storage Temperature: Refrigerator
CAS Number: 91420-49-4

Order Now

Specification Test Results

Purity Limit 95%
Appearance Pale Yellow Oil


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR