3-azidopropyl 4-methylbenzenesulfonate 95%
Supplier: AFG BIOSCIENCE LLC
C56827-1G
C56827-5G
76620-310EA
311.53
CAD
76620-310
76620-312
3-azidopropyl 4-methylbenzenesulfonate 95%
3-azidopropyl 4-methylbenzenesulfonate
Formula:
C₁₀H₁₃N₃O₃S MW: 255.3 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD28977340 CAS Number: 153207-76-2 |
Specification Test Results
Purity Limit | 95% |
Appearance | White solid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...