1-​azido-​3-​iodopropane 95%

Supplier: AFG BIOSCIENCE LLC

Synonyms: 3-azido-1-iodopropane

C56762-1G C56762-5G
76620-300EA 712.09 CAD
76620-300 76620-302
1-​azido-​3-​iodopropane 95%
1-​azido-​3-​iodopropane
Formula: C₃H₆IN₃
MW: 211 g/mol
Storage Temperature: Refrigerator
CAS Number: 58503-62-1

Order Now

Specification Test Results

Purity Limit 95%
Appearance pale-yellowish oil

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR