5-​azidopentan-​1-​ol 97%

Supplier: AFG BIOSCIENCE LLC

Synonyms: 5-Azido-1-pentanol

C56752-1G C56752-5G
76620-296EA 311.53 CAD
76620-296 76620-298
5-​azidopentan-​1-​ol 97%
5-​azidopentan-​1-​ol
Formula: C₅H₁₁N₃O
MW: 129.16 g/mol
Storage Temperature: Ambient
CAS Number: 170220-11-8

Order Now

Specification Test Results

Purity Limit 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR