3-​azidopropan-​1-​ol

Supplier: AFG BIOSCIENCE LLC

Synonyms: 1-Azidopropan-3-ol

C56710-1G C56710-5G
76620-292EA 272.58 CAD
76620-292 76620-294
3-​azidopropan-​1-​ol
3-​azidopropan-​1-​ol
Formula: C₃H₇N₃O
MW: 101.11 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD14652506
CAS Number: 72320-38-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR