2-​Azidoethan-​1-​amine hydrochloride 95%

Supplier: AFG Bioscience

Synonyms: 2-Azidoethylamine hydrochloride

C56669-1G C56669-2G
76620-278EA 474.73 CAD
76620-278 76620-280
2-​Azidoethan-​1-​amine hydrochloride 95%
2-​Azidoethan-​1-​amine hydrochloride
Formula: C₂H₇ClN₄
Storage Temperature: Ambient
MDL Number: MFCD22196453
CAS Number: 118508-67-1

Order Now

Specification Test Results

Purity Limit 95%
Appearance white solid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR