2-​(2-​(2-​azidoethoxy)​ethoxy)​ethanol 95%

Supplier: AFG Bioscience

C56452-10G C56452-1G C56452-5G
76620-266EA 632.96 CAD
76620-266 76620-262 76620-264
2-​(2-​(2-​azidoethoxy)​ethoxy)​ethanol 95%
2-​(2-​(2-​azidoethoxy)​ethoxy)​ethanol
Formula: C₆H₁₃N₃O₃
Storage Temperature: Refrigerator
MDL Number: MFCD09753542
CAS Number: 86520-52-7

Order Now

Specification Test Results

Purity Limit 95%
Appearance Clear Colorless to Yellow Oil


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR