Fmoc-Val-Cit-N-[(4-​bromomethyl])phenyl]​ ≥95%

Supplier: AFG BIOSCIENCE LLC

C56067-100MG C56067-250MG C56067-500MG
76620-234EA 545.18 CAD
76620-234 76620-236 76620-238
Fmoc-Val-Cit-N-[(4-​bromomethyl])phenyl]​ ≥95%
Fmoc-Val-Cit-N-[(4-​bromomethyl])phenyl]​
Formula: C₃₃H₃₈BrN₅O₅
MW: 664.59 g/mol
Storage Temperature: Freezer
CAS Number: 1350456-74-4

Order Now

Specification Test Results

Purity Limit > 95%
Appearance Off-White Powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR