Boc-​Val-​Cit-​PABC-​PNP 95%

Supplier: AFG Bioscience

C56036-1G C56036-250MG C56036-500MG
76620-232EA 1323.99 CAD
76620-232 76620-228 76620-230
Boc-​Val-​Cit-​PABC-​PNP 95%
Boc-​Val-​Cit-​PABC-​PNP
Formula: C₃₀H₄₀N₆O₁₀
Storage Temperature: Freezer
MDL Number: MFCD22417105
CAS Number: 870487-10-8

Order Now

Specification Test Results

Purity Limit 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR