Fmoc-Val-Cit-OH 95%

Supplier: AFG Bioscience

Synonyms: (S)-2-((S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-methylbutanamido)-5-ureidopentanoic acid

C56026-1G C56026-500MG C56026-5G
76620-212EA 77.88 CAD
76620-212 76620-210 76620-214
Fmoc-Val-Cit-OH 95%
Fmoc-Val-Cit-OH
Formula: C₂₆H₃₂N₄O₆
Storage Temperature: Refrigerator
MDL Number: MFCD28139060
CAS Number: 159858-21-6

Order Now

Specification Test Results

Purity Limit 95%
Appearance white powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR